
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii)

Synonym(s):
Acyl-Neuraminyl Hydrolase, Sialidase
Products
Product Category
Brand
Physical Form
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing