Skip to Content
Merck
5-(Aminosulfonyl)-4-chloro-2-[[(tetrahydro-2-furanyl)methyl]amino]benzoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

5-(Aminosulfonyl)-4-chloro-2-[[(tetrahydro-2-furanyl)methyl]amino]benzoic acid

5-(Aminosulfonyl)-4-chloro-2-[[(tetrahydro-2-furanyl)methyl]amino]benzoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
4-Chloro-5-sulfamoyl-N-(tetrahydrofurfuryl)anthranilic acid
Empirical Formula (Hill Notation):
C12H15ClN2O5S
CAS Number:
Molecular Weight:
334.78

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade