CRISPR/Cas9 Products
Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Product | Product Description | |
Pre-designed CRISPR gRNAs | - Synthetic SygRNA® gRNAs available in two-part (crRNA:tracrRNA) or one-part sgRNA formats. - Perfect for animal model generation and cell model engineering. - Recommended to complex with Cas9 protein. - Lentivirus gRNAs are available as ready-to-use lentivirus transduction particles. - Perfect for difficult-to-transfect or primary cells. - Plasmid DNA transfection-grade plasmid gRNAs available in a variety of vectors. - Perfect for transfection, nucleofection, and lentiviral packaging (Note: lentiviral packaging can only be achieved by selecting a lentiviral vector). - In Vitro Transcribed RNA is provided in DNase-, RNase-, and protease-free molecular-grade water. | Order |
Design CRISPR | - Click on the CRISPR Design Tool to get started. - Select your desired genome, choose your gene of interest, and set parameters for the designs. - View and export a list of ranked gRNAs. | Design CRISPR |
CRISPR gRNAs | - Click on the Order gRNAs button and upload your gRNA sequences. - Choose the format: Synthetic gRNA, Transfection-grade plasmid DNA, Lentiviral transduction particles, or IVT gRNA. - Customize the vector and configuration. - Visit the CRISPR Essentials page to compare options. - Purchase clone quantities to receive additional discounts. | Order |
Sanger Whole Genome Knockout (KO) Arrayed CRISPR Libraries | - Over 74,000 individual off-the-shelf CRISPR gRNAs designed to maximize KO of every protein-coding gene in human and mouse genomes. - Available as individual clones, gene panels, and whole genome libraries. | |
Sanger QuickPick™ KO gRNAs | - A fast, cost-efficient way to quickly knock out your gene of interest. - Available formats: Bacteria Glycerol Stocks, Plasmid DNA, Lentiviral transduction particles | Order |
Whole Genome KO Pooled Libraries | - GeCKO v2 CRISPR Knockout Pooled Libraries - Available for human and mouse genomes - Vector options: lentiCRISPRv2 (Cas9+gRNA on same vector); lentiGuide-Puro (gRNA-only vector) |