ATEH03AB2047
AstaTech, Inc.
2-(2-HYDROXYETHOXY)ETHYL BENZOATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-(2-hydroxyethoxy)ethyl benzoate
Inchi Code
1S/C11H14O4/c12-6-7-14-8-9-15-11(13)10-4-2-1-3-5-10/h1-5,12H,6-9H2
InChI key
DNUPYEDSAQDUSO-UHFFFAOYSA-N
Purity
95%
Physical Form
Pale-yellow to Yellow-brown Liquid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=113045
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=A11016
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service