Merck
  • Effect of fiber blends, total solids, heat treatment, whey protein concentrate and stage of sugar incorporation on dietary fiber-fortified
Effect of fiber blends, total solids, heat treatment, whey protein concentrate and stage of sugar incorporation on dietary fiber-fortified
Journal of food science and technology (2017-10-21)
Simran Kaur Arora, A A Patel
ABSTRACT
Owing to the proven beneficial role of dietary fiber (DF) on human health, feasibility of incorporating commercially available soluble and insoluble DF preparations into rice-milk-pudding (
Product Description
Supelco
Total Dietary Fiber Assay Kit, sufficient for ~100 assays