Merck
  • A paraptosis-like cell death induced by δ-tocotrienol in human colon carcinoma SW620 cells is associated with the suppression of the Wnt signaling pathway.
A paraptosis-like cell death induced by δ-tocotrienol in human colon carcinoma SW620 cells is associated with the suppression of the Wnt signaling pathway.
Toxicology (2011-04-02)
Jing-Shu Zhang, Da-Ming Li, Ning He, Ying-Hua Liu, Chun-Hua Wang, Shu-Qing Jiang, Bing-Qing Chen, Jia-Ren Liu
ABSTRACT
Tocotrienol is considered a beneficial effect agent on inhibition of tumor development. In this study, we focused on the effects of δ-tocotrienol and its possible mechanism on induction of death in human colon cancer SW620 cells. δ-Tocotrienol inhibited proliferation of SW620 cell in a dose-dependent manner. Our findings showed that δ-tocotrienol effectively induced paraptosis-like death in SW620 cells, correlated with the vacuolation that may be from welling and fusion of mitochondria and/or the endoplasmic reticulum (ER) as well as caspase-3 nonactivated. However, there were no changes in apoptosis based on flow cytometry analysis. Of being noted, δ-tocotrienol reduced the expression of β-catenin and wnt-1 proteins by about 50% at the highest dose (20μmol/L). δ-Tocotrienol also decreased cyclin D1, c-jun and MMP-7 protein levels in SW620 cells. Altogether, these data indicate that δ-tocotrienol induces paraptosis-like cell death, which is associated with the suppression of the Wnt signaling pathway. Thus, our findings may provide a novel application in treatment of human colon carcinoma.
Product Description
Supelco
δ-Tocotrienol, analytical standard
Sigma-Aldrich
D-δ-Tocotrienol, from rice, ≥65% (GC)