Merck
  • Scaffold-directed and traceless synthesis of tricyclic quinoxalinone imidazoles under microwave irradiation: for SCS-09 special issue-combinatorial approaches to drug discovery.
Scaffold-directed and traceless synthesis of tricyclic quinoxalinone imidazoles under microwave irradiation: for SCS-09 special issue-combinatorial approaches to drug discovery.
Molecular diversity (2010-03-20)
Suman Thummanagoti, Chih-Hau Chen, Zhan-Hui Xu, Chung-Ming Sun
ABSTRACT
Traceless synthesis of 2-aminoimidazoquinoxalinones has been performed on soluble polymer support under open-vessel microwave dielectric heating. The reaction progression is monitored directly by the conventional proton NMR which indicated no release of the substrate from the support. Fmoc-deprotected amino acid polymer conjugates react with 1,5-difluoro-2,4-dinitro benzene to yield polymer bound dinitro fluoro amines, which are further substituted by various primary amines to yield PEG-immobilized dinitrodiamines. Simultaneous reduction of aromatic meta-dinitro group leads to the traceless release of 2-quinoxalinones, followed by N-hetero cyclization with various isothiocyanates in the presence of mercury(II)chloride to furnish 2-aminoimidazoquinolinone rings with three points of diversity at rapid pace.
Sigma-Aldrich
1,5-Difluoro-2,4-dinitrobenzene, 97%