Merck
  • Modification of wood with isopropyl glycidyl ether and its effects on decay resistance and light stability.
Modification of wood with isopropyl glycidyl ether and its effects on decay resistance and light stability.
Bioresource technology (2005-07-26)
Hui-Ting Chang, Shang-Tzen Chang
ABSTRACT
China fir (Cunninghamia lanceolata var. lanceolata) and maple (Acer sp.) wood were etherified with isopropyl glycidyl ether and the decay resistance and light stability of the modified wood were assessed. CP/MAS (13)C NMR and FT-IR analyses indicated that new ether bonds containing isopropyl groups formed after reacting wood with isopropyl glycidyl ether. Modified wood samples were very resistant to decay when exposed to brown-rot fungus Laetiporus sulphureus or white-rot fungus Lenzites betulina for 60 days in the soil-block test. The isopropyl glycidyl ether treatment of wood was effective in decreasing formation of phenoxyl radicals upon UV irradiation and thus protecting wood from photodiscoloration.
Sigma-Aldrich
Glycidyl isopropyl ether, 98%