Merck
  • Synthesis and Characterization of Alkyne-Functionalized Photo-Cross-Linkable Polyesters.
Synthesis and Characterization of Alkyne-Functionalized Photo-Cross-Linkable Polyesters.
ACS omega (2022-05-17)
Warrick Ma, Xiaochu Ding, Ying Chen, Yadong Wang
ABSTRACT
An alkyne-functionalized elastomer derived from sebacic acid, 1,3-propanediol, and alkyne-functionalized serinol is synthesized via melt condensation. A low-power UV lamp triggers the cross-linking rapidly via thiol-yne click chemistry. The cross-linking behavior is studied by photorheology and NMR spectroscopy. The resultant elastomer possesses mechanical properties similar to those of human soft tissues and exhibits in vitro degradability and good cytocompatibility.
Sigma-Aldrich
Human Umbilical Artery Endothelial Cells: HUAEC, neonatal