Merck
Adsorption of 1-Monoglycerides at the Hexane/Water Interface.
Journal of colloid and interface science (1999-12-23)
Hayami, Motomura
ABSTRACT
The interfacial tension of a hexane solution of 1-monolaurin against water was measured as a function of temperature and concentration under atmospheric pressure. The thermodynamic quantity changes associated with the adsorption of 1-monolaurin were evaluated and compared with those of the previously reported 1-monomyristin. The decrease of two carbon atoms in the hydrocarbon chain results in a slight expansion of the 1-monolaurin adsorbed film and in a slight decrease in entropy and energy changes compared with those of the 1-monomyristin system. The large negative value of the entropy change at a high concentration is related to the restricted orientation of the polar head group of 1-monolaurin at the hexane/water interface due to the strong interaction between the large hydrophilic group of 1-monolaurin and the water molecules, as in the 1-monomyristin system. The origin of the distinction in the entropy change behavior between the adsorption from the hexane phase and water phase was discussed. The usefulness of an easier calculation process for the partial molar entropy change is verified by comparison with the usual reliable value and with the entropy of adsorption. Copyright 1999 Academic Press.
Sigma-Aldrich
rac-Glycerol 1-myristate, ≥99%