Merck
High level antibodies to TORCH in the IVIG preparation from Taiwanese.
Journal of the Chinese Medical Association : JCMA (2019-06-11)
Ya-Ling Chou, Kao-Hsian Hsieh, Cherng-Lih Perng, Hueng-Chuen Fan, Chiung-Hsi Tien, Chih-Chien Wang, Shyi-Jou Chen, Fung-Wei Chang
ABSTRACT
Congenital TORCH (toxoplasmosis, other viruses [varicella-zoster virus, VZV, etc.], rubella, cytomegalovirus [CMV], Herpes simplex virus [HSV]) infections are major causes of prenatal, perinatal, and postnatal morbidity and mortality. Although treatment or prevention strategies are available for these pathogens, all drugs may not be safe during the pregnancy. The aim of this study is to measure the antibodies (Abs) concentration in the intravenous immunoglobulin (IVIG) preparation to evaluate the therapeutic potential for TORCH infection. We tested the only one commercial IVIG preparation from Taiwanese for the presence of Abs against Toxoplasma gondii, VZV, Epstein-Barr virus (EBV), measles, mumps, rubella, CMV, HSV type 1 (HSV-1), and HSV type 2 (HSV-2) by using enzyme-linked immunosorbent assay or chemiluminescent microparticle immunoassay. In our study, the median level (range) of anti-CMV immunoglobulin G (IgG) is > 250 (All > 250) (arbitrary unit, AU)/mL, anti-EBV > 200 (All > 200) (relative unit, RU)/mL, anti-HSV > 200 (152.75 to >200) RU/mL, anti-VZV > 5000 (All > 5000) IU/L, anti-measles > 5000 (All > 5000) IU/L, anti-mumps > 200 (156.5 to > 200) RU/mL, anti-rubella 209.8 IU/mL (192.7 to 238.5), and anti-Toxoplasma is 14.05 (12.3 to 16) IU/mL. There was not any immunoglobulin M (IgM) against HSV, VZV, mumps, measles, rubella, CMV, EBV, and Toxoplasma in the "Taiwan Blood Services Foundation" IVIG preparations. There was high activity against T. gondii, VZV, EBV, measles, mumps, rubella, CMV, HSV-1, and HSV-2 in all IVIG batches. Further investigation is warranted to confirm the efficacy of IVIG from Taiwanese for congenital TORCH infections.