
Anti-BTK antibody produced in rabbit

Synonym(s):
Anti-ATK antibody produced in rabbit, Anti-Agammaglobulinemia tyrosine kinase antibody produced in rabbit, Anti-B cell progenitor kinase antibody produced in rabbit, Anti-BPK antibody produced in rabbit, Anti-Bruton tyrosine kinase antibody produced in rabbit, Anti-Tyrosine-protein kinase BTK antibody produced in rabbit, Aγglobulinaemia tyrosine kinase, AGMX1, ATK, B cell progenitor kinase, BPK, Anti-ATK, Anti-BPK
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing