
Acetylsalicylic acid for peak identification

Synonym(s):
Acetylsalicylic acid, 2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin
Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing