
4-Nitrophenyl α-L-rhamnopyranoside

Synonym(s):
p-Nitrophenyl 6-deoxy-α-L-mannopyranoside
Empirical Formula (Hill Notation):
C12H15NO7
CAS Number:
Molecular Weight:
285.25
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing