Merck
  • Central Administration of 5Z-7-Oxozeaenol Protects Experimental Autoimmune Encephalomyelitis Mice by Inhibiting Microglia Activation.
Central Administration of 5Z-7-Oxozeaenol Protects Experimental Autoimmune Encephalomyelitis Mice by Inhibiting Microglia Activation.
Frontiers in pharmacology (2017-11-23)
Lingli Lu, Xiuping Zhang, Huichun Tong, Wenlong Zhang, Pingyi Xu, Shaogang Qu
RÉSUMÉ
Transforming growth factor β-activated kinase 1 (TAK1), a vital upstream integrator of multiple pro-inflammatory signaling pathways, mediates the production of pro-inflammatory cytokines, chemokines, and adhesion molecules. Investigations targeting TAK1 provide new therapeutic options for chronic inflammatory disorders, autoimmune diseases, and cancer. However, the role and mechanism of the TAK1 inhibitor 5Z-7-oxozeaenol in treating autoimmune demyelinating diseases remain unclear. This work aimed to identify whether 5Z-7-oxozeaenol exerts neuroprotective effects on experimental autoimmune encephalomyelitis (EAE) in mice. Here, we demonstrate that 5Z-7-oxozeaenol efficiently alleviates the symptoms of EAE by decreasing the levels of pro-inflammatory cytokines in splenocytes and central nervous system, diminishing the number of activated microglia and inhibiting the p38MAPK, JNK, and ERK signaling pathways. Furthermore, we demonstrate that administration during the symptomatic time window is required for 5Z-7-oxozeaenol efficacy. These results suggest that TAK1 inhibition may provide a potent approach toward treating autoimmune demyelinating diseases.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Adjuvant incomplet de Freund, liquid
Sigma-Aldrich
5Z-7-Oxozeaenol, ≥98% (HPLC)