Merck
Interannual variation of water isotopologues at Vostok indicates a contribution from stratospheric water vapor.
Proceedings of the National Academy of Sciences of the United States of America (2013-06-27)
Renato Winkler, Amaelle Landais, Camille Risi, Melanie Baroni, Alexey Ekaykin, Jean Jouzel, Jean Robert Petit, Frederic Prie, Benedicte Minster, Sonia Falourd
RÉSUMÉ
Combined measurements of water isotopologues of a snow pit at Vostok over the past 60 y reveal a unique signature that cannot be explained only by climatic features as usually done. Comparisons of the data using a general circulation model and a simpler isotopic distillation model reveal a stratospheric signature in the (17)O-excess record at Vostok. Our data and theoretical considerations indicate that mass-independent fractionation imprints the isotopic signature of stratospheric water vapor, which may allow for a distinction between stratospheric and tropospheric influences at remote East Antarctic sites.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Beryllium, chunks, 99.9%