Merck
  • Determination of quinolone residues in infant and young children powdered milk combining solid-phase extraction and ultra-performance liquid chromatography-tandem mass spectrometry.
Determination of quinolone residues in infant and young children powdered milk combining solid-phase extraction and ultra-performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2011-06-21)
Antonio V Herrera-Herrera, Javier Hernández-Borges, Miguel A Rodríguez-Delgado, Miguel Herrero, Alejandro Cifuentes
RÉSUMÉ
The present work describes a method based on solid-phase extraction (SPE) and ultra-performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS) for the simultaneous determination of three quinolones (pipemidic acid, oxolinic acid and flumequine) and twelve fluoroquinolones (marbofloxacin, fleroxacin, pefloxacin, levofloxacin, norfloxacin, ciprofloxacin, enrofloxacin, danofloxacin, lomefloxacin, difloxacin, sarafloxacin, and moxifloxacin) in different infant and young children powdered milks. After suitable deproteination of the reconstituted powdered samples, a SPE procedure was developed providing recovery values higher than 84% (RSDs lower than 13%) for all the analytes, with limits of detection between 0.04 and 0.52 μg/kg. UPLC-MS/MS analyses were carried out in less than 10 min. Sixteen infant and young children powdered milk samples of different origin, type and composition bought at Spanish markets were analyzed. Residues of the selected antibiotics were not detected in any of the analyzed samples.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Oxolinic acid, quinolone antibiotic
Supelco
Oxolinic acid, analytical standard