CLS3840M
Pyrex® extraction apparatus with Allihn condenser, jointed
capacity 250 mL
Synonyme(s) :
Corning® 3840
About This Item
Joint
female joint ST/NS extractor45/50
male joint ST/NS extractor24/40
Fabricant/nom de marque
Corning 3840-M
Capacité
250 mL
Catégories apparentées
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.