Merck
  • Mechanism for the Coupled Photochemistry of Ammonia and Acetylene: Implications for Giant Planets, Comets and Interstellar Organic Synthesis.
Mechanism for the Coupled Photochemistry of Ammonia and Acetylene: Implications for Giant Planets, Comets and Interstellar Organic Synthesis.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (2017-08-10)
Thomas C Keane
ABSTRACT
Laboratory studies provide a fundamental understanding of photochemical processes in planetary atmospheres. Photochemical reactions taking place on giant planets like Jupiter and possibly comets and the interstellar medium are the subject of this research. Reaction pathways are proposed for the coupled photochemistry of NH
Sigma-Aldrich
Hydrazine, anhydrous, 98%
Sigma-Aldrich
Acetaldehyde ammonia trimer, ≥96.0% (NT)