Merck
Growth kinetics of plasma-polymerized films.
Scientific reports (2015-06-19)
Sukyoung Hwang, Hosung Seo, Dong-Cheol Jeong, Long Wen, Jeon Geon Han, Changsik Song, Yunseok Kim
ABSTRACT
The growth kinetics of polymer thin films prepared by plasma-based deposition method were explored using atomic force microscopy. The growth behavior of the first layer of the polythiophene somewhat differs from that of the other layers because the first layer is directly deposited on the substrate, whereas the other layers are deposited on the polymer itself. After the deposition of the first layer, each layer is formed with a cycle of 15 s. The present work represents the growth kinetics of the plasma-polymerized films and could be helpful for further studies on growth kinetics in other material systems as well as for applications of plasma-polymerized thin films.
Sigma-Aldrich
Thiophene, ≥99%