Merck
  • Carbon-supported palladium and ruthenium nanoparticles: application as catalysts in alcohol oxidation, cross-coupling and hydrogenation reactions.
Carbon-supported palladium and ruthenium nanoparticles: application as catalysts in alcohol oxidation, cross-coupling and hydrogenation reactions.
Recent patents on nanotechnology (2012-09-06)
Eduardo J García-Suárez, Patricia Lara, Ana B García, Karine Philippot
ABSTRACT
In the last fifteen-years, the application of metal nanoparticles as catalysts in organic synthesis has received a renewed interest. Therefore, much attention is currently being paid to the synthesis of metal nanoparticles in order to achieve the control of their characteristics in terms of size, shape and surface chemistry. Besides this, the recyclability as well as the recovery from the reaction medium still remain the major drawbacks to widespread the use of nanoparticles in catalysis. To overcome these problems, the immobilization of metal nanoparticles on solid supports appears as a promising alternative. In that context, carbon materials offer several advantages as solid supports such as availability, relatively low cost, high mechanical strength, chemical stability, and a pore structure along with an attractive surface chemistry which allows easy modifications, such as its functionalization, to suit the nanoparticles immobilization needs. Among the transition metals Palladium and Ruthenium are widely employed as efficient catalysts in many reactions. Herein, the most recent advances, from recent papers and patents, in relation to the preparation of carbon-supported Pd or Ru nanoparticles systems as well as their application as catalysts in alcohol oxidation, cross-coupling or hydrogenation reactions, are reviewed.
Sigma-Aldrich
Ruthenium black
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ruthenium, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.75%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 6mm disks, thickness 0.03mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 15mm disks, thickness 0.075mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 0.1m coil, thickness 0.1mm, coil width 2mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 4mm disks, thickness 0.50mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 25mm disks, thickness 0.008mm, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 8mm disks, thickness 0.025mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 4mm disks, thickness 0.03mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Palladium, foil, 8mm disks, thickness 0.047mm, as rolled, 99.95%