Merck
  • Transient non-cardiogenic pulmonary edema following massive ingestion of ethylene glycol butyl ether.
Transient non-cardiogenic pulmonary edema following massive ingestion of ethylene glycol butyl ether.
Intensive care medicine (1992-01-01)
P Bauer, M Weber, J M Mur, J C Protois, P E Bollaert, A Condi, A Larcan, H Lambert
ABSTRACT
A case of acute poisoning with ethylene glycol butyl ether (EGBE) is reported in a chronic alcohol abuser. On admission the 53-year-old patient was comatose with metabolic acidosis, shock, and noncardiogenic pulmonary edema confirmed by haemodynamic study. Following supportive treatment and haemodialysis the outcome was favorable. The relationship between respiratory failure and EGBE is examined.
Sigma-Aldrich
Ethylene glycol butyl ether, SAJ first grade, ≥99.0%
Supelco
Ethylene glycol butyl ether, analytical standard
Sigma-Aldrich
Ethylene glycol butyl ether, ≥99%
Sigma-Aldrich
Ethylene glycol butyl ether, spectrophotometric grade, ≥99.0%