Merck
  • Ibuprofen pharmacokinetics: use of liquid chromatography with radial compression separation.
Ibuprofen pharmacokinetics: use of liquid chromatography with radial compression separation.
Arzneimittel-Forschung (1983-01-01)
D J Greenblatt, R M Arendt, A Locniskar
ABSTRACT
The non-steroidal antiinflammatory agent ibuprofen (Motrin) can be reliably quantitated in body fluids using high-pressure liquid chromatography with a newly developed radial compression separation system. After addition of the structural analogue flurbiprofen as internal standard, biological samples (plasma or cerebrospinal fluid) are acidified and extracted directly into benzene: isoamyl alcohol (98.5:1.5). The organic extract is separated, evaporated to dryness, reconstituted, and chromatographed using a mobile phase of water: acetonitrile (50:50, with 2.5 ml/l acetic acid), a reverse-phase C-18 radial compression cartridge, and UV detection at 229 nm. The routine sensitivity limit is 0.5 micrograms of ibuprofen per ml of sample: this can be improved with minor modifications. The coefficient of variation for replicate samples is less than 7%. Using an automated sampling system, up to 100 samples can be analyzed per 24-hour period. Application of the method to experimental and clinical pharmacokinetic studies of ibuprofen is illustrated.
Product Description
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)