Merck
  • Influence of antimicrobial additives on the formation of rosin nanofibers via electrospinning.
Influence of antimicrobial additives on the formation of rosin nanofibers via electrospinning.
Colloids and surfaces. B, Biointerfaces (2013-01-22)
R Nirmala, Baek Woo-il, R Navamathavan, Duraisamy Kalpana, Yang Soo Lee, Hak Yong Kim
ABSTRACT
In this study, we describe the influence of antimicrobial additives on the formation of rosin fibers by using electrospinning technique. Systematic experiments were performed to fabricate the rosin fibers via electrospinning and we tried to reduce the size of the fibers by mixing some additives such as triethylbenzylammonium chloride (TEBAC), chitosan and silver nitrate in the rosin polymer solution. The morphology, structure and thermal properties of the electrospun rosin fibers were characterized by scanning electron microscopy (SEM), X-ray diffraction (XRD), Fourier-transform infrared spectroscopy (FT-IR), ultraviolet-visible spectroscopy and thermogravimetry (TGA). Rosin fibers with a diameter of the order of nanoscale were achieved by the use of TEBAC additive. The antimicrobial activity of the resultant fibers was checked by the antimicrobial disc diffusion test. All the rosin fibers showed excellent antibacterial activity against the gram negative bacteria and feeble activity against the gram positive bacteria. The present study demonstrated that the electrospun rosin fibers can be utilized for potential antimicrobial products.
Sigma-Aldrich
Gum rosin, natural resin
Sigma-Aldrich
Gum rosin, CP