Merck
  • Spray water reactivation/pelletization of spent CaO-based sorbent from calcium looping cycles.
Spray water reactivation/pelletization of spent CaO-based sorbent from calcium looping cycles.
Environmental science & technology (2012-10-24)
Vasilije Manovic, Yinghai Wu, Ian He, Edward J Anthony
ABSTRACT
This paper presents a novel method for reactivation of spent CaO-based sorbents from calcium looping (CaL) cycles for CO(2) capture. A spent Cadomin limestone-derived sorbent sample from a pilot-scale fluidized bed (FBC) CaL reactor is used for reactivation. The calcined sorbent is sprayed by water in a pelletization vessel. This reactivation method produces pellets ready to be used in FBC reactors. Moreover, this procedure enables the addition of calcium aluminate cement to further enhance sorbent strength. The characterization of reactivated material by nitrogen physisorption (BET, BJH) and scanning electron microscopy (SEM) confirmed the enhanced morphology of sorbent particles for reaction with CO(2). This improved CO(2) carrying capacity was demonstrated in calcination/carbonation tests performed in a thermogravimetric analyzer (TGA). Finally, the resulting pellets displayed a high resistance to attrition during fluidization in a bubbling bed.
Sigma-Aldrich
Calcium oxide, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Calcium oxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium oxide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Calcium oxide, nanopowder, <160 nm particle size (BET), 98%
Sigma-Aldrich
Calcium oxide, reagent grade