Merck
  • Simultaneous determination of 117 pesticides and 30 mycotoxins in raw coffee, without clean-up, by LC-ESI-MS/MS analysis.
Simultaneous determination of 117 pesticides and 30 mycotoxins in raw coffee, without clean-up, by LC-ESI-MS/MS analysis.
Analytica chimica acta (2018-01-14)
Bárbara Reichert, André de Kok, Ionara Regina Pizzutti, Jos Scholten, Carmem Dickow Cardoso, Martien Spanjer
ABSTRACT
This paper describes the optimization and validation of an acetonitrile based method for simultaneous extraction of multiple pesticides and mycotoxins from raw coffee beans followed by LC-ESI-MS/MS determination. Before extraction, the raw coffee samples were milled and then slurried with water. The slurried samples were spiked with two separate standard solutions, one containing 131 pesticides and a second with 35 mycotoxins, which were divided into 3 groups of different relative concentration levels. Optimization of the QuEChERS approach included performance tests with acetonitrile acidified with acetic acid or formic acid, with or without buffer and with or without clean-up of the extracts before LC-ESI-MS/MS analysis. For the clean-up step, seven d-SPE sorbents and their various mixtures were evaluated. After method optimization a complete validation study was carried out to ensure adequate performance of the extraction and chromatographic methods. The samples were spiked at 3 concentrations levels with both mycotoxins and pesticides (with 6 replicates at each level, n = 6) and then submitted to the extraction procedure. Before LC-ESI-MS/MS analysis, the acetonitrile extracts were diluted 2-fold with methanol, in order to improve the chromatographic performance of the early-eluting polar analytes. Calibration standard solutions were prepared in organic solvent and in blank coffee extract at 7 concentration levels and analyzed 6 times each. The method was assessed for accuracy (recovery %), precision (RSD%), selectivity, linearity (r
Product Description
Supelco
Quinalphos, PESTANAL®, analytical standard
Pricing and availability is not currently available.