- Paxillin Binding to the Cytoplasmic Domain of CD103 Promotes Cell Adhesion and Effector Functions for CD8
Paxillin Binding to the Cytoplasmic Domain of CD103 Promotes Cell Adhesion and Effector Functions for CD8
Cancer research (2017-10-13)
Ludiane Gauthier, Stéphanie Corgnac, Marie Boutet, Gwendoline Gros, Pierre Validire, Georges Bismuth, Fathia Mami-Chouaib
PMID29021139
ABSTRACT
CD8