Merck
  • Intracisternal nor-binaltorphimine distinguishes central and peripheral kappa-opioid antinociception in rhesus monkeys.
Intracisternal nor-binaltorphimine distinguishes central and peripheral kappa-opioid antinociception in rhesus monkeys.
The Journal of pharmacology and experimental therapeutics (1999-11-24)
M C Ko, M D Johnson, E R Butelman, K J Willmont, H I Mosberg, J H Woods
ABSTRACT
Systemic administration of nor-binaltorphimine (nor-BNI) produces a long-lasting kappa-opioid receptor (kappaOR) antagonism and has kappa(1)-selectivity in nonhuman primates. The aim of this study was to establish the pharmacological basis of central kappaOR antagonism in rhesus monkeys (Macaca mulatta). After intracisternal (i.c.) administration of small doses of nor-BNI, the duration and selectivity of nor-BNI antagonism were evaluated against two kappaOR agonists, (trans)-3, 4-dichloro-N-methyl-N-[2-(1-pyrrolidinyl)-cyclohexyl]benzeneacetamide (U50,488) and bremazocine. Thermal antinociception was measured in the warm water (50 degrees C) tail-withdrawal assay and sedation was evaluated by observers blind to treatment conditions. Following i.c. pretreatment with 0.32 mg nor-BNI, a 5- to 10-fold rightward shift of the U50,488 baseline dose-effect curve was observed in antinociception. In contrast, this dose of nor-BNI only produced an insignificant 2-fold shift against bremazocine. Pretreatment with a smaller dose (0.032 mg) of nor-BNI produced a 3-fold shift of U50, 488, which lasted for 7 days, but failed to alter the potency of bremazocine. This differential antagonism profile of i.c. nor-BNI also was observed in sedation ratings. In addition, the centrally effective dose of nor-BNI (0.32 mg), when administered s.c. in the back, did not antagonize either U50,488- or bremazocine-induced antinociception and sedation. After i.c. pretreatment with the same dose, nor-BNI also did not antagonize the peripherally mediated effect of U50,488 against capsaicin-induced thermal nociception in the tail. These results indicate that i.c. nor-BNI produces central kappaOR antagonism and support the notion of two functional kappaOR subtypes in the central nervous system. Moreover, it provides a valuable pharmacological basis for further characterizing different sources of kappaOR-mediated effects, namely, from central or peripheral nervous system receptors.