- Flocculation of
Flocculation of
Frontiers in plant science (2017-12-07)
Jianhua Fan, Lvhong Zheng, Yunpeng Bai, Shai Saroussi, Arthur R Grossman
PMID29209355
ABSTRACT
Concentrating algal cells by flocculation as a prelude to centrifugation could significantly reduce the energy and cost of harvesting the algae. However, how variation in phenotypic traits such as cell surface features, cell size and motility alter the efficiency of metal cation and pH-induced flocculation is not well understood. Our results demonstrate that both wild-type and cell wall-deficient strains of the green unicellular alga