Merck
  • DC-SIGN reacts with TLR-4 and regulates inflammatory cytokine expression via NF-κB activation in renal tubular epithelial cells during acute renal injury.
DC-SIGN reacts with TLR-4 and regulates inflammatory cytokine expression via NF-κB activation in renal tubular epithelial cells during acute renal injury.
Clinical and experimental immunology (2017-09-13)
D Feng, Y Wang, Y Liu, L Wu, X Li, Y Chen, Y Chen, Y Chen, C Xu, K Yang, T Zhou
ABSTRACT
In the pathological process of acute kidney injury (AKI), innate immune receptors are essential in inflammatory response modulation; however, the precise molecular mechanisms are still unclear. Our study sought to demonstrate the inflammatory response mechanisms in renal tubular epithelial cells via Toll-like receptor-4 (TLR-4) and dendritic cell-specific intercellular adhesion molecule 3-grabbing non-integrin 1 (DC-SIGN) signalling. We found that DC-SIGN exhibited strong expression in renal tubular epithelial cells of human acute renal injury tissues. DC-SIGN protein expression was increased significantly when renal tubular epithelial cells were exposed to lipopolysaccharide (LPS) for a short period. Furthermore, DC-SIGN was involved in the activation of p65 by TLR-4, which excluded p38 and c-Jun N-terminal kinases (JNK). Interleukin (IL)-6 and tumour necrosis factor (TNF)-α expression was decreased after DC-SIGN knock-down, and LPS induced endogenous interactions and plasma membrane co-expression between TLR-4 and DC-SIGN. These results show that DC-SIGN and TLR-4 interactions regulate inflammatory responses in renal tubular epithelial cells and participate in AKI pathogenesis.