- Aluminum-Doped Cesium Lead Bromide Perovskite Nanocrystals with Stable Blue Photoluminescence Used for Display Backlight.
Aluminum-Doped Cesium Lead Bromide Perovskite Nanocrystals with Stable Blue Photoluminescence Used for Display Backlight.
Bright and stable blue emitters with narrow full-width at half-maxima are particularly desirable for applications in television displays and related technologies. Here, this study shows that doping aluminum (Al