Merck
  • Insecticide susceptibility of Aedes albopictus and Ae. aegypti from Brazil and the Swiss-Italian border region.
Insecticide susceptibility of Aedes albopictus and Ae. aegypti from Brazil and the Swiss-Italian border region.
Parasites & vectors (2017-09-21)
Tobias Suter, Mônica Maria Crespo, Mariana Francelino de Oliveira, Thaynan Sama Alves de Oliveira, Maria Alice Varjal de Melo-Santos, Cláudia Maria Fontes de Oliveira, Constância Flávia Junqueira Ayres, Rosângela Maria Rodrigues Barbosa, Ana Paula Araújo, Lêda Narcisa Regis, Eleonora Flacio, Lukas Engeler, Pie Müller, Maria Helena Neves Lobo Silva-Filha
ABSTRACT
Aedes aegypti and Ae. albopictus are two highly invasive mosquito species, both vectors of several viruses, including dengue, chikungunya and Zika. While Ae. aegypti is the primary vector in the tropics and sub-tropics, Ae. albopictus is increasingly under the public health watch as it has been implicated in arbovirus-transmission in more temperate regions, including continental Europe. Vector control using insecticides is the pillar of most control programmes; hence development of insecticide resistance is of great concern. As part of a Brazilian-Swiss Joint Research Programme we set out to assess whether there are any signs of existing or incipient insecticide resistance primarily against the larvicide Bacillus thuringiensis svar. israelensis (Bti), but also against currently applied and potentially alternative insecticides in our areas, Recife (Brazil) and the Swiss-Italian border region. Following World Health Organization guidelines, dose-response curves for a range of insecticides were established for both colonized and field caught Ae. aegypti and Ae. albopictus. The larvicides included Bti, two of its toxins, Cry11Aa and Cry4Ba, Lysinibacillus sphaericus, Vectomax CG®, a formulated combination of Bti and L. sphaericus, and diflubenzuron. In addition to the larvicides, the Swiss-Italian Ae. albopictus populations were also tested against five adulticides (bendiocarb, dichlorodiphenyltrichloroethane, malathion, permethrin and λ-cyhalothrin). Showing a similar dose-response, all mosquito populations were fully susceptible to the larvicides tested and, in particular, to Bti which is currently used both in Brazil and Switzerland. In addition, there were no signs of incipient resistance against Bti as larvae were equally susceptible to the individual toxins, Cry11Aa and Cry4Ba. The field-caught Swiss-Italian populations were susceptible to the adulticides tested but DDT mortality rates showed signs of reduced susceptibility. The insecticides currently used for mosquito control in Switzerland and Brazil are still effective against the target populations. The present study provides an important reference as relatively few insecticide susceptibility surveys have been carried out with Ae. albopictus.
Product Description
Supelco
Diflubenzuron, PESTANAL®, analytical standard
Pricing and availability is not currently available.