- The Effect of Moderate Temperatures on Latent Fingerprint Chemistry.
The Effect of Moderate Temperatures on Latent Fingerprint Chemistry.
Applied spectroscopy (2017-09-02)
Andrew Johnston, Keith Rogers
PMID28862035
ABSTRACT
The effect of moderate temperatures (25-75 ℃) on latent fingerprints over a five-hour period was examined using Fourier transform infrared (FT-IR) microspectroscopy. The aim of the study was to detect changes in IR spectra due to any changes in fingerprint chemistry; these results were then compared to pure compounds found in sebum that was subjected to 75 ℃ for 5 h. Latent fingerprints deposited on CaF