Merck
  • Determination of polycyclic aromatic hydrocarbons, dibenzothiophene, and alkylated homologs in the lichen Hypogymnia physodes by gas chromatography using single quadrupole mass spectrometry and time-of-flight mass spectrometry.
Determination of polycyclic aromatic hydrocarbons, dibenzothiophene, and alkylated homologs in the lichen Hypogymnia physodes by gas chromatography using single quadrupole mass spectrometry and time-of-flight mass spectrometry.
Journal of chromatography. A (2017-03-08)
William B Studabaker, Keith J Puckett, Kevin E Percy, Matthew S Landis
ABSTRACT
Development of the Athabasca Oil Sands Region in northeastern Alberta, Canada has contributed polycyclic aromatic hydrocarbons (PAHs) and polycyclic aromatic compounds (PACs), which include alkyl PAHs and dibenzothiophenes, to the regional environment. A new analytical method was developed for quantification of PAHs and PACs in the epiphytic lichen bioindicator species Hypogymnia physodes for use in the development of receptor models for attribution of PAH and PAC concentrations to anthropogenic and natural emission sources. Milled lichens were extracted with cyclohexane, and extracts were cleaned on silica gel using automated solid phase extraction techniques. Quantitative analysis was performed by gas chromatography with selected ion monitoring (GC-SIM-MS) for PAHs, and by GC with time-of-flight mass spectrometry (GC-TOF-MS) for PACs. PACs were quantitated in groups using representative reference compounds as calibration standards. Analytical detection limits were ≤2.5ngg
Product Description
1-Methylbenz[a]anthracene, BCR®, certified reference material
Sigma-Aldrich
7-Methylbenzo[a]pyrene, 98%