Merck
Rapidly inducible Cas9 and DSB-ddPCR to probe editing kinetics.
Nature methods (2017-07-25)
John C Rose, Jason J Stephany, William J Valente, Bridget M Trevillian, Ha V Dang, Jason H Bielas, Dustin J Maly, Douglas M Fowler
ABSTRACT
We developed a chemically inducible Cas9 (ciCas9) and a droplet digital PCR assay for double-strand breaks (DSB-ddPCR) to investigate the kinetics of Cas9-mediated generation and repair of DSBs in cells. ciCas9 is a rapidly activated, single-component Cas9 variant engineered by replacing the protein's REC2 domain with the BCL-xL protein and fusing an interacting BH3 peptide to the C terminus. ciCas9 can be tunably activated by a compound that disrupts the BCL-xL-BH3 interaction within minutes. DSB-ddPCR demonstrates time-resolved, highly quantitative, and targeted measurement of DSBs. Combining these tools facilitated an unprecedented exploration of the kinetics of Cas9-mediated DNA cleavage and repair. We find that sgRNAs targeting different sites generally induce cleavage within minutes and repair within 1 or 2 h. However, we observe distinct kinetic profiles, even for proximal sites, and this suggests that target sequence and chromatin state modulate cleavage and repair kinetics.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Pricing and availability is not currently available.
Sigma-Aldrich
Boron-11B, 95 atom % 11B
Pricing and availability is not currently available.