Merck
  • Novel Chromatographic Methods for Simultaneous Quantification of Fish and Wheat Germ Oils Mixture in Pharmaceutical Dosage Forms.
Novel Chromatographic Methods for Simultaneous Quantification of Fish and Wheat Germ Oils Mixture in Pharmaceutical Dosage Forms.
Journal of chromatographic science (2017-02-17)
Amira F El-Yazbi, Amr El-Hawiet
ABSTRACT
Two simple, direct and environment-friendly chromatographic methods, high-performance liquid chromatography (HPLC) and high-performance thin layer chromatographic (HPTLC), were developed for the determination of a binary mixture of fish oil (FO) and wheat germ oil (WGO), for the first time, in their pharmaceutical dosage forms with no need for any sample pretreatment. The HPLC separation was carried out using C-18 stationary phase with mobile phase of 15% formic acid (pH 6), methanol and acetonitrile through gradient-elution, 1.5 mL min-1 flow-rate and detection at 215 nm for FO and 280 nm for WGO. HPTLC separation was carried out on silica-coated plates using diethyl ether-petroleum ether (0.5:9.5, v/v) as mobile phase. Detection was at 215 nm for FO and 240 nm for WGO. Regression analysis showed good linear relationship with r > 0.999 in the concentration-ranges of 0.2-2 mg mL-1 and 2.5-20 μg band-1 for WGO by HPLC and HPTLC methods, respectively, and 0.4-10 mg mL-1 and 25-200 μg band-1 for FO by HPLC and HPTLC methods, respectively. The methods were validated, showed good analytical performance and were successfully applied for the analysis of pharmaceutical formulations and synthetic mixtures of the analytes with good recoveries. Therefore, the two methods could be conveniently adopted for routine analysis of similar products in quality control laboratories of pharmaceutical industries especially that simultaneous determination of FO-WGO mixture has not been reported previously.