Merck
  • Chirality and energy transfer amplified circularly polarized luminescence in composite nanohelix.
Chirality and energy transfer amplified circularly polarized luminescence in composite nanohelix.
Nature communications (2017-06-07)
Dong Yang, Pengfei Duan, Li Zhang, Minghua Liu
ABSTRACT
Transfer of both chirality and energy information plays an important role in biological systems. Here we show a chiral donor π-gelator and assembled it with an achiral π-acceptor to see how chirality and energy can be transferred in a composite donor-acceptor system. It is found that the individual chiral gelator can self-assemble into nanohelix. In the presence of the achiral acceptor, the self-assembly can also proceed and lead to the formation of the composite nanohelix. In the composite nanohelix, an energy transfer is realized. Interestingly, in the composite nanohelix, the achiral acceptor can both capture the supramolecular chirality and collect the circularly polarized energy from the chiral donor, showing both supramolecular chirality and energy transfer amplified circularly polarized luminescence (ETACPL).
Sigma-Aldrich
9,10-Bis(phenylethynyl)anthracene, 97%
Pricing and availability is not currently available.