- Discharge of KPC-2 genes from the WWTPs contributed to their enriched abundance in the receiving river.
Discharge of KPC-2 genes from the WWTPs contributed to their enriched abundance in the receiving river.
The Science of the total environment (2017-01-10)
Fengxia Yang, Liang Huang, Linyun Li, Yang Yang, Daqing Mao, Yi Luo
PMID28065546
ABSTRACT
At present, very little is known about the persistence and spread pathway of KPC-2 genes in the environment. Our previous study reported the prevalence and persistence of KPC-2 genes in wastewater treatment plants (WWTPs). In the present work, we investigated the occurrence and fate of KPC-2 genes in a WWTP discharge-receiving river and studied the effect of WWTP discharges on the prevalence of KPC-2 genes and host bacteria in the receiving river. It is observed that a considerable level of KPC-2 genes occurred in the receiving river, and a significant increase of bla
MATERIALS