- Development of pro-inflammatory phenotype in monocytes after engulfing Hb-activated platelets in hemolytic disorders.
Development of pro-inflammatory phenotype in monocytes after engulfing Hb-activated platelets in hemolytic disorders.
Clinical immunology (Orlando, Fla.) (2017-01-01)
Rashi Singhal, Sheetal Chawla, Deepak K Rathore, Angika Bhasym, Gowtham K Annarapu, Vandana Sharma, Tulika Seth, Prasenjit Guchhait
PMID28039017
ABSTRACT
Monocytes and macrophage combat infections and maintain homeostatic balance by engulfing microbes and apoptotic cells, and releasing inflammatory cytokines. Studies have described that these cells develop anti-inflammatory properties upon recycling the free-hemoglobin (Hb) in hemolytic conditions. While investigating the phenotype of monocytes in two hemolytic disorders-paroxysmal nocturnal hemoglobinuria (PNH) and sickle cell disease (SCD), we observed a high number of pro-inflammatory (CD14