Merck
Effects of intra-hippocampal microinjection of vitamin B
Physiology & behavior (2016-12-22)
Amir Erfanparast, Esmaeal Tamaddonfard, Shaghayegh Nemati
ABSTRACT
In the present study, we investigated the effects of microinjection of vitamin B
Sigma-Aldrich
Naloxonazine dihydrochloride hydrate, powder, ≥95% (HPLC)