- Seasonal variations and source profile of n-alkanes in particulate matter (PM
Seasonal variations and source profile of n-alkanes in particulate matter (PM
Delhi is one of the most polluted cities in the world. The generation of aerosols in the lower atmosphere of the city is mainly due to a large amount of natural dust advection and sizable anthropogenic activities. The compositions of organic compounds in aerosols are highly variable in this region and need to be investigated thoroughly. Twenty-four-hour sampling to assess concentrations of n-alkanes (ng/m