Merck
Methylation of Notch3 modulates chemoresistance via P-glycoprotein.
European journal of pharmacology (2016-11-05)
Xiaoting Gu, Yangfan Lu, Dongxu He, Chunxiao Lu, Jian Jin, Xiaojie Lu, Xin Ma
ABSTRACT
The global gene expression and DNA methylation of genes in adriamycin-resistant human breast cancer cells (MCF-7/ADM cells) are similar to those in paclitaxel-resistant MCF-7 cells (MCF-7/PTX) and are significantly different from those in wild-type MCF-7 cells. DNA methylation is associated with chemoresistance in breast cancer and changes the characteristics of chemoresistant and chemosensitive cells. Here, we showed that the tumor-suppressor gene Notch3 was inactivated due to epigenetic silencing DNA hypermethylation in MCF-7/ADM cells. In addition, the drug efflux pump P-glycoprotein was negatively regulated by Notch3 and highly expressed in MCF-7/ADM cells. Taken together, our findings demonstrated that hypermethylation of Notch3 causes activation of P-glycoprotein in adriamycin-resistant cells.
Sigma-Aldrich
α,α-Dicyanoethyl acetate, 98%