Merck
Estrogen receptor-α in medial amygdala neurons regulates body weight.
The Journal of clinical investigation (2015-06-23)
Pingwen Xu, Xuehong Cao, Yanlin He, Liangru Zhu, Yongjie Yang, Kenji Saito, Chunmei Wang, Xiaofeng Yan, Antentor Othrell Hinton, Fang Zou, Hongfang Ding, Yan Xia, Chunling Yan, Gang Shu, San-Pin Wu, Bin Yang, Yuxin Feng, Deborah J Clegg, Richard DeMarchi, Sohaib A Khan, Sophia Y Tsai, Francesco J DeMayo, Qi Wu, Qingchun Tong, Yong Xu
ABSTRACT
Estrogen receptor-α (ERα) activity in the brain prevents obesity in both males and females. However, the ERα-expressing neural populations that regulate body weight remain to be fully elucidated. Here we showed that single-minded-1 (SIM1) neurons in the medial amygdala (MeA) express abundant levels of ERα. Specific deletion of the gene encoding ERα (Esr1) from SIM1 neurons, which are mostly within the MeA, caused hypoactivity and obesity in both male and female mice fed with regular chow, increased susceptibility to diet-induced obesity (DIO) in males but not in females, and blunted the body weight-lowering effects of a glucagon-like peptide-1-estrogen (GLP-1-estrogen) conjugate. Furthermore, selective adeno-associated virus-mediated deletion of Esr1 in the MeA of adult male mice produced a rapid body weight gain that was associated with remarkable reductions in physical activity but did not alter food intake. Conversely, overexpression of ERα in the MeA markedly reduced the severity of DIO in male mice. Finally, an ERα agonist depolarized MeA SIM1 neurons and increased their firing rate, and designer receptors exclusively activated by designer drug-mediated (DREADD-mediated) activation of these neurons increased physical activity in mice. Collectively, our results support a model where ERα signals activate MeA neurons to stimulate physical activity, which in turn prevents body weight gain.
Sigma-Aldrich
Anti-Glutamate antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Monoclonal Anti-GABA antibody produced in mouse, clone GB-69, ascites fluid