Merck
  • The Expression of VHL (Von Hippel-Lindau) After Traumatic Spinal Cord Injury and Its Role in Neuronal Apoptosis.
The Expression of VHL (Von Hippel-Lindau) After Traumatic Spinal Cord Injury and Its Role in Neuronal Apoptosis.
Neurochemical research (2016-06-22)
Jie Hao, Xiaoqing Chen, Ting Fu, Jie Liu, Mingchen Yu, Wei Han, Shuang He, Rong Qian, Feng Zhang
ABSTRACT
The VHL (Von Hippel-Lindau) gene is a tumor suppressor gene, which is best known as an E3 ubiquitin ligase that negatively regulates the hypoxia inducible factor. The inactivation of VHL gene could result in the abnormal synthesis of VHL protein, which is in contact with the development and occurrence of renal clear cell carcinoma. However, the expression and possible function of VHL in central nervous system (CNS) is still unclear. To examine the function of VHL in CNS injury and repair, we used an acute spinal cord injury (SCI) model in adult rats. Western blot analysis showed an important upregulation of VHL protein, reaching a peak at day 3 and then declined during the following days. Double immunofluorescence staining showed that VHL was co-expressed with neurons, but not with astrocytes and microglia. Moreover, we detected that active caspase-3 had co-localized with VHL in neurons after SCI. Additionally in vitro, VHL depletion, by short interfering RNA, significantly reduced neuronal apoptosis. In conclusion, these data suggested that the change of VHL protein expression was related to neuronal apoptosis after SCI.