Merck
  • Efficient synthesis of substituted vinyl ethers using the Julia olefination.
Efficient synthesis of substituted vinyl ethers using the Julia olefination.
Organic letters (2003-12-05)
Simon Surprenant, Wing Yan Chan, Carl Berthelette
ABSTRACT
Julia olefination between alpha-alkoxy sulfones 2a-c and a wide variety of ketones or aldehydes afforded substituted vinyl ethers in 46-90% yields. Sulfones 2a-c were readily prepared in two steps from commercially available reagents in 68-80% yields. Optimization revealed that the nature of the base, the solvent, and the temperature were crucial to obtaining the desired vinyl ethers. [reaction: see text]
Sigma-Aldrich
Benzo[b]thiophene-2-carboxaldehyde, 97%