Merck
  • Discovery and structure-activity relationship of 3-aryl-5-aryl-1,2,4-oxadiazoles as a new series of apoptosis inducers and potential anticancer agents.
Discovery and structure-activity relationship of 3-aryl-5-aryl-1,2,4-oxadiazoles as a new series of apoptosis inducers and potential anticancer agents.
Journal of medicinal chemistry (2005-08-05)
Han-Zhong Zhang, Shailaja Kasibhatla, Jared Kuemmerle, William Kemnitzer, Kristin Ollis-Mason, Ling Qiu, Candace Crogan-Grundy, Ben Tseng, John Drewe, Sui Xiong Cai
ABSTRACT
We have identified 5-(3-chlorothiophen-2-yl)-3-(4-trifluoromethylphenyl)-1,2,4-oxadiazole (1d) as a novel apoptosis inducer through our caspase- and cell-based high-throughput screening assay. Compound 1d has good activity against several breast and colorectal cancer cell lines but is inactive against several other cancer cell lines. In a flow cytometry assay, treatment of T47D cells with 1d resulted in arrest of cells in the G(1) phase, followed by induction of apoptosis. SAR studies of 1d showed that the 3-phenyl group can be replaced by a pyridyl group, and a substituted five-member ring in the 5-position is important for activity. 5-(3-Chlorothiophen-2-yl)-3-(5-chloropyridin-2-yl)-1,2,4-oxadiazole (4l) has been found to have in vivo activity in a MX-1 tumor model. Using a photoaffinity agent, the molecular target has been identified as TIP47, an IGF II receptor binding protein. Therefore, our cell-based chemical genetics approach for the discovery of apoptosis inducers can identify potential anticancer agents as well as their molecular targets.
Sigma-Aldrich
4-(Trifluoromethyl)benzamidoxime, 97%