Merck
  • Distinguishing between Leucine and Isoleucine by Integrated LC-MS Analysis Using an Orbitrap Fusion Mass Spectrometer.
Distinguishing between Leucine and Isoleucine by Integrated LC-MS Analysis Using an Orbitrap Fusion Mass Spectrometer.
Analytical chemistry (2016-11-02)
Yongsheng Xiao, Malgorzata M Vecchi, Dingyi Wen
ABSTRACT
Despite the great success of mass spectrometry (MS) for de novo protein sequencing, Leu and Ile have been generally considered to be indistinguishable by MS because their molecular masses are exactly the same. Positioning of incorrect Leu/Ile residues in variable domains, especially in CDRs (complementarity determining regions) of an antibody, may result in substantial loss of antigen binding affinity and specificity of the antibody. Here, we describe an integrated LC-MS based strategy, encompassing a combination of HCD (high-energy collisional dissociation) multistage mass spectrometric analysis (HCD-MS