Merck
  • Postsynthetic modification of peptoids via the Suzuki-Miyaura cross-coupling reaction.
Postsynthetic modification of peptoids via the Suzuki-Miyaura cross-coupling reaction.
Biopolymers (2015-09-26)
Ho Yeon Nam, Jiwon Seo
ABSTRACT
We developed a new method for modifying the side chains of peptoids on a solid phase resin, employing the palladium-catalyzed Suzuki-Miyaura cross-coupling reaction. Optimized conditions using Pd(PPh3 )4 and K2 CO3 in the presence of Buchwald's SPhos ligand provided a high conversion in the coupling reaction. The usefulness of this method was demonstrated by synthesis of a two pyrene-conjugated peptoid helix, which exhibited an interesting excimer formation.
Sigma-Aldrich
4-Iodobenzylamine, AldrichCPR
Sigma-Aldrich
4-Bromobenzylamine, 96%