Merck
FOXO3a-Dependent Mechanism of E1A-Induced Chemosensitization.
Cancer research (2011-09-14)
Jen-Liang Su, Xiaoyun Cheng, Hirohito Yamaguchi, Yi-Wen Chang, Chao-Feng Hou, Dung-Fang Lee, How-Wen Ko, Kuo-Tai Hua, Ying-Nai Wang, Michael Hsiao, Poshen B Chen, Jung-Mao Hsu, Robert C Bast, Gabriel N Hortobagyi, Mien-Chie Hung
ABSTRACT
Gene therapy trials in human breast, ovarian, and head and neck tumors indicate that adenovirus E1A can sensitize cancer cells to the cytotoxic effects of paclitaxel in vitro and in vivo. Resistance to paclitaxel has been reported to occur in cells expressing low levels of the Forkhead transcription factor FOXO3a. In this article, we report that FOXO3a is critical for E1A-mediated chemosensitization to paclitaxel. RNA interference-mediated knockdown of FOXO3a abolished E1A-induced sensitivity to paclitaxel. Mechanistic investigations indicated that E1A indirectly stabilized FOXO3a by acting at an intermediate step to inhibit a ubiquitin-dependent proteolysis pathway involving the E3 ligase βTrCP and the FOXO3a inhibitory kinase IKKβ. E1A derepressed this inhibitory pathway by stimulating expression of the protein phosphatase 2A (PP2A)/C protein phosphatases, which by binding to the TGF-β-activated kinase TAK1, inhibited its ability to activate IKKβ and, thereby, to suppress βTrCP-mediated degradation of FOXO3a. Thus, by stimulating PP2A/C expression, E1A triggers a signaling cascade that stabilizes FOXO3a and mediates chemosensitization. Our findings provide a leap forward in understanding paclitaxel chemosensitization by E1A, and offer a mechanistic rational to apply E1A gene therapy as an adjuvant for improving therapeutic outcomes in patients receiving paclitaxel treatment.