Merck
  • Survival of the replication checkpoint deficient cells requires MUS81-RAD52 function.
Survival of the replication checkpoint deficient cells requires MUS81-RAD52 function.
PLoS genetics (2013-11-10)
Ivana Murfuni, Giorgia Basile, Shyamal Subramanyam, Eva Malacaria, Margherita Bignami, Maria Spies, Annapaola Franchitto, Pietro Pichierri
ABSTRACT
In checkpoint-deficient cells, DNA double-strand breaks (DSBs) are produced during replication by the structure-specific endonuclease MUS81. The mechanism underlying MUS81-dependent cleavage, and the effect on chromosome integrity and viability of checkpoint deficient cells is only partly understood, especially in human cells. Here, we show that MUS81-induced DSBs are specifically triggered by CHK1 inhibition in a manner that is unrelated to the loss of RAD51, and does not involve formation of a RAD51 substrate. Indeed, CHK1 deficiency results in the formation of a RAD52-dependent structure that is cleaved by MUS81. Moreover, in CHK1-deficient cells depletion of RAD52, but not of MUS81, rescues chromosome instability observed after replication fork stalling. However, when RAD52 is down-regulated, recovery from replication stress requires MUS81, and loss of both these proteins results in massive cell death that can be suppressed by RAD51 depletion. Our findings reveal a novel RAD52/MUS81-dependent mechanism that promotes cell viability and genome integrity in checkpoint-deficient cells, and disclose the involvement of MUS81 to multiple processes after replication stress.
Sigma-Aldrich
RAD51 Inhibitor B02, ≥98% (HPLC)
Pricing and availability is not currently available.